1-(2-Ethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71654-5G 71654-1G 71654-25G 71654-500MG
77633-696EA 1432 USD
77633-696 77639-932 77660-282 77651-950
1-(2-Ethylphenyl)-2,2-difluoroethanone ≥95%
1-(2-Ethylphenyl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD20920913
CAS Number: 1352209-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR