2,2-Difluoro-1-(3,4,5-trichlorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71598-5G 71598-1G 71598-500MG
77656-832EA 2245 USD
77656-832 77643-966 77647-928
2,2-Difluoro-1-(3,4,5-trichlorophenyl)ethanone ≥95%
2,2-Difluoro-1-(3,4,5-trichlorophenyl)ethanone

Precaución: For Research Use Only

Formula: 259.465 g/mol
MW: C₈H₃Cl₃F₂O
Storage Temperature: Ambient
MDL Number: MFCD34815480
CAS Number: 2112842-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR