1-(3-(Benzyloxy)-5-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

71571-5G 71571-1G 71571-25G 71571-500MG
77630-624EA 1441 USD
77630-624 77639-930 77660-280 77651-948
1-(3-(Benzyloxy)-5-methylphenyl)ethanol ≥95%
1-(3-(Benzyloxy)-5-methylphenyl)ethanol

Precaución: For Research Use Only

Formula: 242.3129 g/mol
MW: C₁₆H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34764169
CAS Number: 2918864-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR