2-Fluoro-6-methoxy-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

71180-5G 71180-25G 71180-1G 71180-500MG
77630-622EA 1441 USD
77630-622 77656-830 77643-964 77647-922
2-Fluoro-6-methoxy-4-(trifluoromethyl)benzaldehyde ≥97%
2-Fluoro-6-methoxy-4-(trifluoromethyl)benzaldehyde

Precaución: For Research Use Only

Formula: 222.136 g/mol
MW: C₉H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD27975481
CAS Number: 1823920-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR