3,4-Diethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

70814-5G 70814-25G 70814-1G 70814-500MG
77630-620EA 1441 USD
77630-620 77660-274 77643-962 77647-920
3,4-Diethoxy-2-fluorobenzaldehyde ≥97%
3,4-Diethoxy-2-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD18824394
CAS Number: 701263-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR