6-Chloro-2-fluoro-3-formylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

70043-5G 70043-25G 70043-1G 70043-500MG
77633-692EA 712 USD
77633-692 77660-272 77643-960 77651-944
6-Chloro-2-fluoro-3-formylbenzonitrile ≥95%
6-Chloro-2-fluoro-3-formylbenzonitrile

Precaución: For Research Use Only

Formula: 183.567 g/mol
MW: C₈H₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD28363334
CAS Number: 1446401-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR