2-Fluoro-3-formyl-6-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
70041-1G
70041-5G
70041-500MG
77639-924EA
748
USD
77639-924
77656-826
77651-942
2-Fluoro-3-formyl-6-methylbenzoic acid ≥97%
2-Fluoro-3-formyl-6-methylbenzoic acid
Precaución: For Research Use Only
Formula:
182.148 g/mol MW: C₉H₇FO₃ Storage Temperature: Ambient |
MDL Number:
MFCD34815071 CAS Number: 2169353-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...