3-Chloro-2,5-difluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

69868-1G 69868-5G 69868-500MG
77639-922EA 551 USD
77639-922 77660-268 77651-940
3-Chloro-2,5-difluorobenzaldehyde ≥95%
3-Chloro-2,5-difluorobenzaldehyde

Precaución: For Research Use Only

Formula: 176.548 g/mol
MW: C₇H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD18415554
CAS Number: 1261884-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR