2-Fluoro-4-methylisophthalaldehyde ≥95%

Supplier: AOBChem USA

Warning

69742-5G 69742-1G 69742-25G 69742-500MG
77630-618EA 1441 USD
77630-618 77639-920 77660-266 77647-916
2-Fluoro-4-methylisophthalaldehyde ≥95%
2-Fluoro-4-methylisophthalaldehyde

Precaución: For Research Use Only

Formula: C₉H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34764016
CAS Number: 2092334-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR