2,5-Difluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69678-5G 69678-25G 69678-1G 69678-500MG
77630-616EA 1441 USD
77630-616 77660-264 77643-958 77647-914
2,5-Difluoro-3-methylbenzaldehyde ≥97%
2,5-Difluoro-3-methylbenzaldehyde

Precaución: For Research Use Only

Formula: C₈H₆F₂O
Storage Temperature: Ambient
MDL Number: MFCD12025460
CAS Number: 1379096-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR