2,4-difluoro-3-hydroxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

69537-5G 69537-25G 69537-1G 69537-500MG
77633-688EA 771 USD
77633-688 77660-262 77643-956 77647-912
2,4-difluoro-3-hydroxybenzoic acid ≥95%
2,4-difluoro-3-hydroxybenzoic acid

Precaución: For Research Use Only

Formula: C₇H₄F₂O₃
MW: 174.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797839
CAS Number: 91659-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR