1-(2,4-Difluoro-3-(methylthio)phenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

69491-5G 69491-1G 69491-25G 69491-500MG
77633-686EA 1366 USD
77633-686 77639-918 77660-260 77647-910
1-(2,4-Difluoro-3-(methylthio)phenyl)ethanol ≥95%
1-(2,4-Difluoro-3-(methylthio)phenyl)ethanol

Precaución: For Research Use Only

Formula: 204.237 g/mol
MW: C₉H₁₀F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34763953
CAS Number: 1894824-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR