1-(6-Bromo-2-fluoropyridin-3-yl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

69249-5G 69249-25G 69249-1G 69249-500MG
77630-612EA 1441 USD
77630-612 77656-822 77643-954 77647-908
1-(6-Bromo-2-fluoropyridin-3-yl)-2,2-difluoroethanone ≥97%
1-(6-Bromo-2-fluoropyridin-3-yl)-2,2-difluoroethanone

Precaución: For Research Use Only

Formula: C₇H₃BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD32851940
CAS Number: 2918947-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR