2,5-Dibromo-1-chloro-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

69240-5G 69240-25G 69240-1G 69240-500MG
77633-684EA 833 USD
77633-684 77660-256 77643-952 77647-906
2,5-Dibromo-1-chloro-3-methylbenzene ≥95%
2,5-Dibromo-1-chloro-3-methylbenzene

Precaución: For Research Use Only

Formula: 284.376 g/mol
MW: C₇H₅Br₂Cl
Storage Temperature: Ambient
MDL Number: MFCD09878184
CAS Number: 1000573-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR