Ethyl 3-chloro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

69128-5G 69128-1G 69128-500MG
77660-254EA 2001 USD
77660-254 77643-950 77651-938
Ethyl 3-chloro-5-methylbenzoate ≥97%
Ethyl 3-chloro-5-methylbenzoate

Precaución: For Research Use Only

Formula: 198.646 g/mol
MW: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD11554962
CAS Number: 1256479-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR