5'-Chloro-2'-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

69065-5G 69065-1G 69065-25G 69065-500MG
77633-682EA 1061 USD
77633-682 77639-916 77660-250 77647-904
5'-Chloro-2'-fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥95%
5'-Chloro-2'-fluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: For Research Use Only

Formula: 234.653 g/mol
MW: C₁₃H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD16817633
CAS Number: 1242339-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR