1-(2-Bromo-6-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
68996-5G
68996-1G
68996-25G
68996-500MG
77630-610EA
1441
USD
77630-610
77639-914
77656-820
77647-902
1-(2-Bromo-6-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Bromo-6-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone
Precaución: For Research Use Only
Formula:
317.098 g/mol MW: C₉H₅BrF₄OS Storage Temperature: Ambient |
MDL Number:
MFCD35165076 CAS Number: 2756657-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...