4-Bromo-2-fluoro-5-nitrobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

68993-5G 68993-25G
77635-282EA 64 USD
77635-282 77656-818
4-Bromo-2-fluoro-5-nitrobenzaldehyde ≥95%
4-Bromo-2-fluoro-5-nitrobenzaldehyde

Precaución: For Research Use Only

Formula: C₇H₃BrFNO₃
MW: 248.0060 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10758067
CAS Number: 679839-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR