1-(2,5-Dibromopyridin-4-yl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
68935-5G
68935-25G
68935-1G
68935-500MG
77630-608EA
1441
USD
77630-608
77656-816
77643-948
77651-936
1-(2,5-Dibromopyridin-4-yl)-2,2,2-trifluoroethanone ≥95%
1-(2,5-Dibromopyridin-4-yl)-2,2,2-trifluoroethanone
Precaución: For Research Use Only
Formula:
332.9 g/mol MW: C₇H₂Br₂F₃NO Storage Temperature: Ambient |
MDL Number:
MFCD34763701 CAS Number: 2901107-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...