4-Fluoro-3-iodo-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
68905-5G
68905-25G
68905-1G
68905-500MG
77630-606EA
1441
USD
77630-606
77636-198
77643-946
77651-934
4-Fluoro-3-iodo-1,1'-biphenyl ≥97%
4-Fluoro-3-iodo-1,1'-biphenyl
Precaución: For Research Use Only
Formula:
298.095 g/mol MW: C₁₂H₈FI Storage Temperature: Ambient |
MDL Number:
MFCD14701842 CAS Number: 1214328-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...