1-(5-Bromobenzo[d][1,3]dioxol-4-yl)ethanol ≥95%

Supplier: AOBChem USA

Warning

68845-5G 68845-1G 68845-500MG
77630-604EA 1766 USD
77630-604 77643-944 77651-930
1-(5-Bromobenzo[d][1,3]dioxol-4-yl)ethanol ≥95%
1-(5-Bromobenzo[d][1,3]dioxol-4-yl)ethanol

Precaución: For Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD29913479
CAS Number: 249636-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR