1-(2-Bromo-5-chloropyridin-4-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

68842-5G 68842-1G 68842-25G 68842-500MG
77630-602EA 1441 USD
77630-602 77639-912 77660-248 77647-900
1-(2-Bromo-5-chloropyridin-4-yl)-2,2,2-trifluoroethanone ≥95%
1-(2-Bromo-5-chloropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 288.449 g/mol
MW: C₇H₂BrClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD34763666
CAS Number: 2918859-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR