4-Bromo-2-chloro-3-(4-fluorophenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

67935-5G 67935-25G 67935-1G 67935-500MG
77630-600EA 1441 USD
77630-600 77636-196 77639-908 77647-894
4-Bromo-2-chloro-3-(4-fluorophenyl)pyridine ≥95%
4-Bromo-2-chloro-3-(4-fluorophenyl)pyridine

Precaución: For Research Use Only

Formula: 286.527 g/mol
MW: C₁₁H₆BrClFN
Storage Temperature: Ambient
MDL Number: MFCD34763076
CAS Number: 2901096-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR