1-(3,5-Dibromophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67237-5G 67237-25G 67237-1G 67237-500MG
77633-678EA 810 USD
77633-678 77660-240 77643-934 77651-928
1-(3,5-Dibromophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3,5-Dibromophenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: C₈H₃Br₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD27665749
CAS Number: 1190865-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR