2,2,2-Trifluoro-1-(4-iodophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

67025-5G 67025-25G 67025-1G 67025-500MG
77635-274EA 288 USD
77635-274 77660-238 77643-930 77647-884
2,2,2-Trifluoro-1-(4-iodophenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-iodophenyl)ethanone

Precaución: For Research Use Only

Formula: C₈H₄F₃IO
Storage Temperature: Ambient
MDL Number: MFCD02260838
CAS Number: 23516-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR