1-(5-(Benzyloxy)-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

67012-5G 67012-25G 67012-1G 67012-500MG
77630-596EA 1441 USD
77630-596 77656-798 77643-928 77647-882
1-(5-(Benzyloxy)-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(5-(Benzyloxy)-2-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 298.232 g/mol
MW: C₁₅H₁₀F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD29040536
CAS Number: 2386585-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR