1-(3-Chloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

66848-1G 66848-5G 66848-500MG
77639-902EA 560 USD
77639-902 77660-236 77651-924
1-(3-Chloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Chloro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 276.563 g/mol
MW: C₉H₃ClF₆O
Storage Temperature: Ambient
MDL Number: MFCD22688461
CAS Number: 1417461-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR