1-(3-Chloro-4-fluoro-5-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

66753-1G 66753-5G 66753-500MG
77635-272EA 1121 USD
77635-272 77656-794 77651-922
1-(3-Chloro-4-fluoro-5-methylphenyl)ethanone ≥97%
1-(3-Chloro-4-fluoro-5-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 186.611 g/mol
MW: C₉H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD28522254
CAS Number: 176548-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR