(4-Chloro-2-methoxyphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

66699-5G 66699-1G 66699-500MG
77630-592EA 1726 USD
77630-592 77639-900 77647-880
(4-Chloro-2-methoxyphenyl)trimethylsilane ≥95%
(4-Chloro-2-methoxyphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 214.764 g/mol
MW: C₁₀H₁₅ClOSi
Storage Temperature: Ambient
MDL Number: MFCD34687545
CAS Number: 2901098-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR