(2,6-Bis(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥95%
Supplier: AOBChem USA
|
Warning
|
66548-5G
66548-500MG
66548-25G
66548-1G
77633-674EA
1441
USD
77633-674
77653-064
77660-232
77643-924
(2,6-Bis(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥95%
(2,6-Bis(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone
Precaución: For Research Use Only
Formula:
311.223 g/mol MW: C₁₃H₁₁F₆NO Storage Temperature: Ambient |
MDL Number:
MFCD34762654 CAS Number: 2324593-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...