2-(Benzyloxy)-1,5-dibromo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

66504-5G 66504-1G 66504-25G 66504-500MG
77630-590EA 1441 USD
77630-590 77639-898 77656-792 77651-916
2-(Benzyloxy)-1,5-dibromo-3-methylbenzene ≥97%
2-(Benzyloxy)-1,5-dibromo-3-methylbenzene

Precaución: For Research Use Only

Formula: 356.052 g/mol
MW: C₁₄H₁₂Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34814527
CAS Number: 100542-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR