2-(3,4-Dichloro-2-fluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

66378-5G 66378-25G 66378-1G 66378-500MG
77630-588EA 1441 USD
77630-588 77656-790 77643-920 77651-912
2-(3,4-Dichloro-2-fluorophenyl)propan-2-ol ≥95%
2-(3,4-Dichloro-2-fluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₉H₉Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD34762545
CAS Number: 2807446-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR