4-(Difluoromethyl)-1,2-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

66258-5G 66258-1G 66258-25G 66258-500MG
77635-270EA 271 USD
77635-270 77639-896 77656-786 77651-910
4-(Difluoromethyl)-1,2-difluorobenzene ≥97%
4-(Difluoromethyl)-1,2-difluorobenzene

Precaución: For Research Use Only

Formula: C₇H₄F₄
Storage Temperature: Ambient
MDL Number: MFCD14698573
CAS Number: 1214379-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR