2-Methyl-5-(pyridin-4-yl)aniline ≥97%

Supplier: AOBChem USA

Warning

66061-5G 66061-1G 66061-500MG
77630-586EA 1515 USD
77630-586 77639-894 77647-876
2-Methyl-5-(pyridin-4-yl)aniline ≥97%
2-Methyl-5-(pyridin-4-yl)aniline

Precaución: For Research Use Only

Formula: 184.237 g/mol
MW: C₁₂H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD12785836
CAS Number: 761441-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR