2,2,2-Trifluoro-1-(4-fluoro-[1,1'-biphenyl]-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65877-5G 65877-1G 65877-500MG
77656-784EA 2223 USD
77656-784 77643-918 77651-904
2,2,2-Trifluoro-1-(4-fluoro-[1,1'-biphenyl]-3-yl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-fluoro-[1,1'-biphenyl]-3-yl)ethanone

Precaución: For Research Use Only

Formula: C₁₄H₈F₄O
Storage Temperature: Ambient
MDL Number: MFCD26687747
CAS Number: 1516570-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR