2,2,2-Trifluoro-1-(3-phenoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65799-5G 65799-1G 65799-25G 65799-500MG
77630-582EA 1441 USD
77630-582 77639-886 77656-780 77647-872
2,2,2-Trifluoro-1-(3-phenoxyphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(3-phenoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 266.215 g/mol
MW: C₁₄H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD16613962
CAS Number: 67851-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR