Perfluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

65796-5G 65796-25G
77635-268EA 132 USD
77635-268 77660-226
Perfluoroacetophenone ≥97%
Perfluoroacetophenone

Precaución: For Research Use Only

Formula: C₈F₈O
MW: 264.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00216596
CAS Number: 652-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR