2,2,2-Trifluoro-1-(5-isopropyl-4-methoxy-2-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65727-1G 65727-5G 65727-500MG
77639-884EA 638 USD
77639-884 77660-224 77647-870
2,2,2-Trifluoro-1-(5-isopropyl-4-methoxy-2-methylphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(5-isopropyl-4-methoxy-2-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: 260.252 g/mol
MW: C₁₃H₁₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07380817
CAS Number: 1368984-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR