1-(4-Ethoxy-2-fluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65345-5G 65345-25G 65345-1G 65345-500MG
77633-668EA 788 USD
77633-668 77660-222 77643-914 77651-900
1-(4-Ethoxy-2-fluorophenyl)ethanone ≥95%
1-(4-Ethoxy-2-fluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 182.192 g/mol
MW: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD28521106
CAS Number: 851667-22-6
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR