1-(2-Chloro-3-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65196-5G 65196-25G 65196-1G 65196-500MG
77630-578EA 1441 USD
77630-578 77656-774 77643-912 77647-866
1-(2-Chloro-3-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(2-Chloro-3-fluoropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: C₇H₂ClF₄NO
Storage Temperature: Ambient
MDL Number: MFCD34762143
CAS Number: 2901066-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR