2,2,2-Trifluoro-1-(2-methoxypyrimidin-5-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

65076-5G 65076-25G 65076-1G 65076-500MG
77630-576EA 1441 USD
77630-576 77656-772 77643-910 77647-864
2,2,2-Trifluoro-1-(2-methoxypyrimidin-5-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-methoxypyrimidin-5-yl)ethanone

Precaución: For Research Use Only

Formula: 206.1220 g/mol
MW: C₇H₅F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26680939
CAS Number: 1533159-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR