1-Bromo-3-chloro-2-iodo-5-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

64987-5G 64987-1G 64987-25G 64987-500MG
77633-666EA 989 USD
77633-666 77639-880 77660-220 77651-896
1-Bromo-3-chloro-2-iodo-5-methoxybenzene ≥95%
1-Bromo-3-chloro-2-iodo-5-methoxybenzene

Precaución: For Research Use Only

Formula: 347.3754 g/mol
MW: C₇H₅BrClIO
Storage Temperature: Ambient
MDL Number: MFCD34762046
CAS Number: 2386078-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR