4-(5-Chloro-3-fluoropyridin-2-yl)morpholine ≥95%

Supplier: AOBChem USA

Warning

64770-5G 64770-25G
77635-266EA 104 USD
77635-266 77660-216
4-(5-Chloro-3-fluoropyridin-2-yl)morpholine ≥95%
4-(5-Chloro-3-fluoropyridin-2-yl)morpholine

Precaución: For Research Use Only

Formula: C₉H₁₀ClFN₂O
Storage Temperature: Ambient
MDL Number: MFCD09972191
CAS Number: 1020253-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR