1-Phenylcyclopentanol ≥95%

Supplier: AOBChem USA

Warning

64342-5G 64342-25G 64342-1G 64342-500MG
77633-664EA 1027 USD
77633-664 77656-768 77643-904 77651-890
1-Phenylcyclopentanol ≥95%
1-Phenylcyclopentanol

Precaución: For Research Use Only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00060796
CAS Number: 10487-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR