1-(2,5-Diethylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64231-5G 64231-25G 64231-1G 64231-500MG
77630-570EA 1441 USD
77630-570 77660-210 77643-902 77651-886
1-(2,5-Diethylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,5-Diethylphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 230.2262 g/mol
MW: C₁₂H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD24107306
CAS Number: 1531142-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR