2,2,2-Trifluoro-1-(2,4,6-trifluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

64160-5G 64160-25G 64160-1G 64160-500MG
77633-662EA 968 USD
77633-662 77660-208 77643-900 77651-884
2,2,2-Trifluoro-1-(2,4,6-trifluorophenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2,4,6-trifluorophenyl)ethanone

Precaución: For Research Use Only

Formula: 228.0912 g/mol
MW: C₈H₂F₆O
Storage Temperature: Ambient
MDL Number: MFCD12547805
CAS Number: 1092712-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR