5’-Chloro-2’-hydroxy-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

64114-5G 64114-500MG 64114-25G 64114-1G
77633-660EA 768 USD
77633-660 77653-062 77656-764 77643-896
5’-Chloro-2’-hydroxy-2,2,2-trifluoroacetophenone ≥95%
5’-Chloro-2’-hydroxy-2,2,2-trifluoroacetophenone

Precaución: For Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23382016
CAS Number: 870614-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR