2,2,2-Trifluoro-1-(4-fluoro-2-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

64087-5G 64087-25G 64087-1G 64087-500MG
77630-566EA 1441 USD
77630-566 77656-762 77643-894 77651-880
2,2,2-Trifluoro-1-(4-fluoro-2-methoxyphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-fluoro-2-methoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 222.1363 g/mol
MW: C₉H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11113544
CAS Number: 1256467-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR