2,2,2-Trifluoro-1-(2-hydroxy-4-methoxyphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64016-500MG
64016-1G
64016-5G
77653-060EA
511
USD
77653-060
77639-872
77656-760
2,2,2-Trifluoro-1-(2-hydroxy-4-methoxyphenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-hydroxy-4-methoxyphenyl)ethanone
Precaución: For Research Use Only
Formula:
C₉H₇F₃O₃ MW: 220.1452 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27958449 CAS Number: 123716-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...