1-(2,5-Dichloro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63896-5G 63896-1G 63896-500MG
77630-562EA 1556 USD
77630-562 77643-892 77647-858
1-(2,5-Dichloro-4-methoxyphenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,5-Dichloro-4-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 273.0360 g/mol
MW: C₉H₅Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07380832
CAS Number: 1368710-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR